Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ogerin

Click to view available options
Quantity:
10 mg
50 mg
Description
Ogerin is a selective positive allosteric modulator (PAM) of GPR68 (pEC50 = 6.83), which exhibits selectivity for GPR68 over 58 other GPCRs. Ogerin attenuates contextual-based fear memory in mice.
Specifications
Specifications
| Chemical Name or Material | 2-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]benzenemethanol |
| CAS | 1309198-71-7 |
| Quantity | 10 mg |
| Target | Orphan 7-TM Receptor Agonists |
| Molecular Formula | C17H17N5O |
| Formula Weight | 307.35 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction