Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGFOD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB049475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049475 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049475 Supplier Novus Biologicals Supplier No. NBP297921AF750

Rabbit Polyclonal Antibody

OGFOD1 Polyclonal specifically detects OGFOD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OGFOD1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.-
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OGFOD1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55239
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.