Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGG1 Antibody (2B4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252724SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252724SS 0.025 mg
NBP252724 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP252724SS Supplier Novus Biologicals Supplier No. NBP252724SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

OGG1 Monoclonal specifically detects OGG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen OGG1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B4
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2-5 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Alias 8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase
Gene Symbols OGG1
Host Species Mouse
Immunogen Partial recombinant protein made to an internal sequence of the human OGG1 protein (between amino acids 100-300) [Uniprot: O15527]
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4968
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.