Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGG1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB100106SS

 View more versions of this product

Catalog No. NB100106SS


Only null left
Add to Cart

Description

Description

OGG1 Polyclonal specifically detects OGG1 in Human, Mouse, Rat, Porcine, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).
Specifications

Specifications

OGG1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase
Rabbit
39 kDa
0.025 mL
Base Excision Repair, Cancer, Chromatin Research, DNA Repair
4968
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Proximity Ligation Assay
1.0 mg/mL
Western Blot 1:500-1:1000, Flow Cytometry reported by customer review, ELISA reported in scientific literature (PMID 19506022), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20956573), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:25-1:100, Immunoblotting reported in multiple pieces of scientific literature, Proximity Ligation Assay reported in scientific literature (PMID 35883652), Flow (Intracellular)
O15527
OGG1
A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527]
Affinity Purified
RUO
Primary
Human, Mouse, Rat, Pig, Primate, Rabbit
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only