Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGG1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$269.00 - $572.50
Specifications
Antigen | OGG1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry reported by customer review, ELISA reported in scientific literature (PMID 19506022), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20956573), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:25-1:100, Immunoblotting reported in multiple pieces of scientific literature, Proximity Ligation Assay reported in scientific literature (PMID 35883652), Flow (Intracellular) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Proximity Ligation Assay |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100106SS
![]() |
Novus Biologicals
NB100106SS |
0.025 mL |
Each for $269.00
|
|
|||||
NB100106
![]() |
Novus Biologicals
NB100106 |
0.1 mL |
Each for $572.50
|
|
|||||
Description
OGG1 Polyclonal specifically detects OGG1 in Human, Mouse, Rat, Porcine, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).Specifications
OGG1 | |
Western Blot 1:500-1:1000, Flow Cytometry reported by customer review, ELISA reported in scientific literature (PMID 19506022), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20956573), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:25-1:100, Immunoblotting reported in multiple pieces of scientific literature, Proximity Ligation Assay reported in scientific literature (PMID 35883652), Flow (Intracellular) | |
Polyclonal | |
Rabbit | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
PBS with 0.05% Sodium Azide | |
8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
OGG1 | |
IgG | |
Affinity Purified | |
39 kDa |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Proximity Ligation Assay | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Pig, Primate, Rabbit | |
O15527 | |
4968 | |
A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title