Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGG1 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB100106V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100106V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100106V Supplier Novus Biologicals Supplier No. NB100106V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OGG1 Polyclonal specifically detects OGG1 in Human, Mouse, Pig, Primate, Rabbit, Rat samples. It is validated for ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot.

Specifications

Antigen OGG1
Applications ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O15527
Gene Alias 8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase
Gene Symbols OGG1
Host Species Rabbit
Immunogen A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527]
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 4968
Target Species Human, Mouse, Pig, Primate, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.