Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B2A6, Proteintech
SDP

Catalog No. 501737213 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7213 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7213 Supplier Proteintech Group Inc Supplier No. 668231IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.

Specifications

Antigen OGT
Applications Western Blot
Classification Monoclonal
Clone 2B2A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OGT
Gene Accession No. O15294, P56558, Q27HV0
Gene Alias FLJ23071, HRNT1, O GLCNAC, OGT
Gene Symbols OGT
Host Species Mouse
Immunogen OGT Fusion Protein Ag28402
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108155, 26295, 664652, 8473
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.