Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B2A6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | OGT |
---|---|
Clone | 2B2A6 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7213
![]() ![]() |
Proteintech Group Inc
668231IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.Specifications
OGT | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FLJ23071, HRNT1, O GLCNAC, OGT | |
OGT | |
IgG2a | |
Protein A | |
Antibody |
2B2A6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
O15294, P56558, Q27HV0 | |
108155, 26295, 664652, 8473 | |
OGT Fusion Protein Ag28402 | |
Primary | |
-20°C | |
OGT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title