Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Olanzapine
GREENER_CHOICE

Catalog No. p-4816662
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT2A/D2 antagonist; atypical antipsychotic

Antagonist of 5-HT2A and dopamine D2 receptors (Ki values are 4 and 11 nM respectively). Also displays affinity for a range of other receptors including D1 and D4, 5-HT2C, α1, H1 and M1-4 receptors (Ki values range between 1.9 and 31 nM) . Atypical antipsychotic. Displays anticholinergic properties. Also a highly potent activator of hM4Di DREADDs (EC50 values are 5 nM in vitro and 7 nM in vivo).

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17H20N4S

Specifications

Quantity 10 mg
Formula Weight 312.43
Purity >99%
Molecular Weight (g/mol) 312.44
Chemical Name or Material Olanzapine
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.