Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OLDA
GREENER_CHOICE

Catalog No. p-4785276
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective endogenous TRPV1 agonist

Potent endogenous vanilloid TRPV1 (VR1) receptor agonist (EC50 = 36 nM at hVR1) with low affinity for rCB1 receptors (Ki = 1.6 μM). Potently induces VR1-mediated thermal hyperalgesia in rats in vivo.

Chemical Identifiers

CAS 105955-11-1
Molecular Formula C26H43NO3

Specifications

Quantity 5 mg
Formula Weight 417.63
Purity >99%
Molecular Weight (g/mol) 417.634
Chemical Name or Material OLDA
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.