Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olfactory Marker Protein Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$192.47 - $529.95
Specifications
| Antigen | Olfactory Marker Protein |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 5-10 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018007
![]() |
Novus Biologicals
NB110747530.1MG |
0.1 mg |
Each for $192.47
|
|
|||||
NB11074753
![]() |
Novus Biologicals
NB11074753 |
0.5 mg |
Each for $529.95
|
|
|||||
Description
Olfactory Marker Protein Polyclonal specifically detects Olfactory Marker Protein in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Olfactory Marker Protein | |
| Western Blot 5-10 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat, Mouse | |
| olfactory marker protein, Olfactory neuronal-specific protein | |
| OMP | |
| Primary | |
| Highly specific for mature olfactory neurons (including axon and terminals). |
| LYOPH | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Olfactory | |
| P08523 | |
| 4975 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title