Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olfactory Marker Protein Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018007 0.1 mg
NB11074753 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018007 Supplier Novus Biologicals Supplier No. NB110747530.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Olfactory Marker Protein Polyclonal specifically detects Olfactory Marker Protein in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Olfactory Marker Protein
Applications Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 5-10 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation PBS with No Preservative
Gene Accession No. P08523
Gene Alias olfactory marker protein, Olfactory neuronal-specific protein
Gene Symbols OMP
Host Species Rabbit
Immunogen A synthetic peptide as a part of rat Olfactory Marker Protein (OMP) conjugated to immunogenic carrier protein has been used as the immunogen. The peptide is homologous with the corresponding sequence derived from OMP protein in mouse.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Olfactory
Primary or Secondary Primary
Gene ID (Entrez) 4975
Test Specificity Highly specific for mature olfactory neurons (including axon and terminals).
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.