Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFM4 Monoclonal Antibody (12)
GREENER_CHOICE

Catalog No. PIMA529457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529457 Supplier Invitrogen™ Supplier No. MA529457
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human OLFM4/GW112.

OLFM4 is a 510 amino acid extracellular matrix glycoprotein belonging to the olfactomedin-related protein family and found to be selectively expressed in inflammed colonic epithelium. It has one signal peptide, six N-linked glycosylation motif and forms disulfide-bonded multimers. OLFM4 a transcription factor may have a matrix-related function in differentiation. Constitutively expressed OLFM4 in cancer cells binds to the potent apoptosis inducer GRIM-19 and has a novel antiapoptotic action and hence might serve as a diagnostic marker for human cancers. Reports suggest that OLFM4 promotes proliferation of PANC-1 cells by favoring transition from the S to G2/M phase. Potential interaction of OLFM4 with endogenous cell surface lectins and cadherins may mediate cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFM4
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% trehalose and no preservative
Gene OLFM4
Gene Accession No. Q6UX06
Gene Alias Antiapoptotic protein GW112; bA209J19.1; GC1; G-CSF-stimulated clone 1 protein; Gm296; Gm913; GW112; hGC-1; hOLfD; olfactomedin 4; olfactomedin-4; OlfD; OLFM4; OLM4; pPD4; PU.1 difference product 4; UNQ362; UNQ362/PRO698
Gene Symbols OLFM4
Host Species Mouse
Immunogen Recombinant Human OLFM4/GW112 protein (Met1-Gln510).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10562
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.