Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFML2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566528
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA566528 100 μL
1 options

Catalog No. PIPA566528

Supplier: Invitrogen™ PA566528

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKAKVSEEEND IEEQQDEFFS GDNGVDLLIE DQLLRHNGLM TSVTRRPAAT RQGHSTAVTS DLNARTAPWS SAL Highest antigen sequence identity to the following orthologs - mouse 69%, rat 64%.

This gene encodes an olfactomedin domain-containing protein. Most olfactomedin domain-containing proteins are secreted glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFML2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OLFML2B
Gene Accession No. Q68BL8
Gene Alias 1110018N05Rik; 4832415H08Rik; AI467542; olfactomedin like 2B; olfactomedin-like 2B; olfactomedin-like protein 2B; Olfml2b; Photomedin-2
Gene Symbols OLFML2B
Host Species Rabbit
Immunogen Recombinant Human OLFML2B. Recombinant protein control fragment (Product #RP-105241).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.