Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig2 Antibody (064), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB087665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087665 Supplier Novus Biologicals Supplier No. NBP289201JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Olig2 Monoclonal specifically detects Olig2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Olig2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 64
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM Sodium Borate
Gene Alias BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the N-terminus of the Human Olig2.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10215
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.