Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP128667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128667 0.1 mL
NBP128667SS 0.02 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP128667 Supplier Novus Biologicals Supplier No. NBP128667
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

Olig2 Polyclonal specifically detects Olig2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Olig2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:3000, Immunohistochemistry 1:800, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:500-1:1000, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen
Formulation 200 mM Tris (pH 8.0), 0.1 M Glycine with No Preservative
Gene Accession No. Q9EQW6
Gene Alias BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1
Gene Symbols OLIG2
Host Species Rabbit
Immunogen Recombinant mouse Olig2. Accession # Q9EQW6
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10215
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.