Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig2 Antibody (OLIG2/2400), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB383359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383359 Supplier Novus Biologicals Supplier No. NBP308960FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Olig2 Monoclonal antibody specifically detects Olig2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Olig2
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone OLIG2/2400
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1
Host Species Mouse
Immunogen Recombinant fragment of human Olig2 protein (around aa 1-141) (exact sequence is proprietary) (Uniprot: Q13516)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10215
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.