Description
- For use in lessons on Bowen's reaction series
- First mineral to crystallize in cooling magma
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- Massive earthy habit is useful example of nonmetallic luster
Specifications
Specifications
| Product Type | Mineral |
| For Use With (Application) | Study of mineral physical properties like hardness, fracture, and luster |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Olivine |