Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLLAS Epitope Tag Antibody (L2), HRP, Novus Biologicals™
SDP

Catalog No. NBP106713H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106713H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106713H Supplier Novus Biologicals Supplier No. NBP106713H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

OLLAS Epitope Tag Monoclonal antibody specifically detects OLLAS Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount.

Specifications

Antigen OLLAS Epitope Tag
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone L2
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount
Gene Alias E.coli OmpF Linker and mouse Langerin fusion Sequence, OLLA-2
Host Species Rat
Immunogen OLLAS (SGFANELGPRLMGK)-tagged extracellular domain of mouse Langerin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Tag specific.
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.