Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLT-2 Antibody (2564C), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB315421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315421 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315421 Supplier Novus Biologicals Supplier No. FAB10059PECY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

OLT-2 Monoclonal specifically detects OLT-2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen OLT-2
Applications Flow Cytometry
Classification Monoclonal
Clone 2564C
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias OSCAR-Like Transcript-2 Protein, T Cell-Interacting, Activating Receptor On Myeloid Cells, T Cell-Interacting, Activating Receptor On Myeloid Cells 1, TARM1, T-Cell-Interacting, Activating Receptor On Myeloid Cells Protein 1
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human OLT-2/TARM-1, Met1-Val235, Accession # B6A8C7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441864
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.