Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OMgp Antibody, Novus Biologicals™
SDP

Catalog No. NB1002557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002557 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002557 Supplier Novus Biologicals Supplier No. NB1002557
Add to Cart
Add to Cart

Goat Polyclonal Antibody

OMgp Polyclonal specifically detects OMgp in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen OMgp
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002535.3
Gene Alias oligodendrocyte myelin glycoprotein, OMGPoligodendrocyte-myelin glycoprotein
Gene Symbols OMG
Host Species Goat
Immunogen Peptide with sequence C-KVTKIPKQYRTKE corresponding to internal region according to NP_002535.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4974
Test Specificity Cross reacts with Human. Expected from sequence similarity: Rat and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.