Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diba Omnifit™ Bottle Waste Cap Thread

Catalog No. p-4398562
Click to view available options
Format:
Four Luer Port

Connect a vapor trap using the vent port to prevent solvent vapors from escaping

  • Use without any fittings or use male Luer fittings to connect various tubing sizes
  • Anti-twist design

Specifications

Product Type Threaded Closure
Pre-sterilized No
Closure Size 38-430 (GL38)
Liner Material PTFE
Autoclavable Autoclavable
Material Polypropylene
Color Blue
For Use With (Application) Waste disposal
Format Four Luer Port
Height (Metric) 2.54 cm
Quantity 1 Each
Size 38-430 (GL38)
Length (Metric) 5.08 cm
Sterility Non-sterile
For Use With (Equipment) Diba Solvent Waste Bottles
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.