Learn More
BD ONCLARITY™ HPV Test
Catalog No. B441990 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
Catalog No. Form
B441990 Ready-to-Use (RTU)
1 options
This item is not returnable. View return policy
Catalog No. B441990 Supplier BD Supplier No. 441990
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
ONCLARITY™ HPV Test for the FDA Approved HPV assay for extended genotyping
  • BD ONCLARITY™ HPV Test for the FDA Approved HPV assay for extended genotyping
  • Extended genotyping supports risk stratification and persistence monitoring to guide patient management
  • Genotypes 16 and 18 account for 70% of invasive cancer worldwide, but their prevalence is declining as vaccination rates increase
  • Genotypes 31, 33, 58 have a CIN3+ risk similar to genotype 18, but 51, 35, 39, 68, 56, 59, 66 have a much lower risk
TRUSTED_SUSTAINABILITY
Specifications
Type HPV Test
Form Ready-to-Use (RTU)