Learn More
Thermo Scientific™ Solid tumor: AcroMetrix Oncology Hotspot Control (IVD, CE-IVD)
GSA_VA
Catalog No. 23279193 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 x 25 μL
Catalog No. Quantity
23-279-193 3 x 25 μL
1 options
Catalog No. 23-279-193 Supplier Thermo Scientific™ Supplier No. 969056
Add to Cart
Add to Cart
Assess day-to-day test variation of somatic mutation panels with the Thermo Scientific™ AcroMetrix™ Oncology Hotspot Control, which is for In Vitro Diagnostic Use.
Assess day-to-day test variation of somatic mutation panels. Intended for use with next generation sequencing (NGS) assays designed to identify somatic mutations in DNA from human samples. For In Vitro Diagnostic Use.
  • Multiple variant types to capture the diversity of somatic cancer mutation types
  • 500+ clinically relevant variants allow detection of multiple targets with NGS in a single reaction
  • IVD labeled enables use in clinical diagnostics
  • Variant frequency near LOD for test method development and validation
FAQs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Oncology Hotspot Control
Packaging 3 x 25µL
Quantity 3 x 25 μL