Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncostatin M/OSM Antibody (015), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB107972 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107972 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107972 Supplier Novus Biologicals Supplier No. NBP289506V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Oncostatin M/OSM
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 15
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias MGC20461, oncostatin M, oncostatin-M
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Oncostatin M/OSM (P13725; Met1-Arg221).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5008
Test Specificity No cross-reactivity in ELISA with: Mouse OSM Human CNTF Human IL2 Human IL6 Human LIF Human GCSF
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.