Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncostatin M/OSM Antibody (17022), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB296601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296601 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296601 Supplier Novus Biologicals Supplier No. FAB2951S

Mouse Monoclonal Antibody

Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOF

Specifications

Antigen Oncostatin M/OSM
Applications Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 17022
Conjugate Alexa Fluor 750
Dilution Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGC20461, oncostatin M, oncostatin-M
Host Species Mouse
Immunogen E. coli-derived recombinant human Oncostatin M/OSM, Ala26-Arg221, Accession # P13725
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5008
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.