Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncostatin M/OSM Antibody (31E7), Novus Biologicals™
SDP

Catalog No. NB298087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298087 100 μL
NB298086 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298087 Supplier Novus Biologicals Supplier No. NBP326445100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA

Specifications

Antigen Oncostatin M/OSM
Applications ELISA
Classification Monoclonal
Clone 31E7
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias MGC20461, oncostatin M, oncostatin-M
Host Species Human
Immunogen Recombinant Human Oncostatin M/OSM protein [UniProt P13725]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5008
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.