Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oncostatin M/OSM Antibody (31E7), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Oncostatin M/OSM |
---|---|
Clone | 31E7 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISASpecifications
Oncostatin M/OSM | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC20461, oncostatin M, oncostatin-M | |
Recombinant Human Oncostatin M/OSM protein [UniProt P13725] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
31E7 | |
ELISA | |
Unconjugated | |
Human | |
Biologically Active Proteins, Cell Cycle and Replication | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
5008 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title