Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncostatin M/OSM Antibody (OTI2B6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273146A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273146A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273146A405 Supplier Novus Biologicals Supplier No. NBP273146AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Oncostatin M/OSM
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B6
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC20461, oncostatin M, oncostatin-M
Gene Symbols OSM
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 25-252 of human Oncostatin M/OSM.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5008.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.