Learn More
Thermo Scientific™ ONE Broth-Salmonella Base
Description
Enrich and recover Salmonella spp. from food samples in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base, when prepared with Thermo Scientific Oxoid ONE Broth-Salmonella Selective Supplement (SR0242).
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Description | ONE Broth-Salmonella Base |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Species | Salmonella |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.