Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ One Shot™ ccdB Survival™ 2 T1R Competent Cells

Catalog No. a10460
Encompass_Preferred
Click to view available options
Quantity:
11 x 50 μL

Includes

ccdB Survival2 T1R Competent Cells: 11 vials, 50μL each, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 1 bottle, 6mL

One Shot ccdB Survival 2 T1R Competent Cells are suitable for the propagation of plasmids containing the ccdB gene and are designed for use with the Gateway Vector Conversion System and for propagating Gateway destination, donor, and supercoiled entry vectors.

One Shot ccdB Survival 2 T1R Competent Cells are suitable for the propagation of plasmids containing the ccdB gene and are designed for use with the Gateway Vector Conversion System and for propagating Gateway destination, donor, and supercoiled entry vectors. ccdB Survival 2 T1R cells that are made chemically competent can achieve high transformation efficiency of >1 x 109 transformants/μg plasmid DNA.

The ccdB Survival 2 T1R E. coli strain is derived from the TOP10 strain and offers the same benefits. Mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) allow cloning of both prokaryotic and eukaryotic genomic DNA that contains methylcytosine and methyladenine. This strain also has the lacZΔM15 genotype, providing the option of blue-white screening on plates containing either X-Gal or Bluo-Gal. In addition, recA1 and endA1 mutations increase insert stability and improve the quality of plasmid DNA prepared from minipreps.The tonA (also known as fhuA) mutation in ccdB Survival 2 T1R cells confers resistance to bacteriophages T1 and T5, safeguarding samples against contamination.

ccdB Survival 2 T1R Competent Cells offer:
• >1 x 109 transformants/μg efficiency
tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• Support of high-yield preparation of supercoiled plasmid DNA (the endA1 phenotype)
• Reduced nonspecific recombination of cloned DNA (the recA1 phenotype) 

Convenient and efficient format
One Shot ccdB Survival 2 T1R E. coli are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation so there are no efficiency-zapping, freeze-thaw cycles or money wasted on unused cells.

Genotype: F-mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG fhuA::IS2

Find the strain and format that fits your needs
• We offer other DH strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• MultiShot formats for high throughput applications are available.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot ccdB Survival 2 T1R Competent Cells (11 x 50 μL)
Store Competent Cells at &ndash80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 ml)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid ccdB vector propagation
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line One Shot™
Quantity 11 x 50 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.