Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVOS™ EVOS™ Onstage Vessel Holder, two 60-mm Petri dishes
Click to view available options
Quantity:
1 Ea.
Description
- Maximizes choice of cell culture vessels when conducting time-lapse imaging experiments using EVOS FL Auto Imaging System equipped with EVOS Onstage Incubator
- Allows perfect fit of micro-well plates or cell culture flasks/dishes to master plate of EVOS Onstage Incubator
Specifications
Specifications
| Type | Vessel Holder |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Auto Imaging System |
| Format | two, 60 mm Petri dishes |
| Holds | 2 60 mm Petri Dishes |
| Quantity | 1 Ea. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction