Learn More
Eisco™ Oolitic Limestone Specimen (Sedimentary Rock)
SCI_ED
Catalog No. s26672
Click to view available options
Quantity:
1 Ea.
Oolitic limestone is a carbonate rock formed from the cementation of thousands of ooids.
  • Composed primarily of ooids
  • Small, smooth pieces of calcium carbonate
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Some samples may contain identifiable fossil material
  • Forms in reef or lagoon environments
  • Useful in the classroom for lessons on depositional facies, sedimentary rocks, and stratigraphy
Specifications
Product Type Metamorphic Rock, Oolitic Limestone
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.