Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPA1 Antibody (1284B) - BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP259770SS
Description
OPA1 Monoclonal specifically detects OPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
OPA1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 | |
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1284B | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
OPA1 | |
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot# O60313] | |
0.025 mg | |
Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision | |
4976 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction