Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPA1 Antibody (1284B) - BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 5 publications
$243.50 - $499.50
Specifications
Antigen | OPA1 |
---|---|
Clone | 1284B |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
NBP259770SS
![]() |
Novus Biologicals
NBP259770SS |
Unconjugated | 0.025 mg |
Each for $243.50
|
|
||||
NBP259770
![]() |
Novus Biologicals
NBP259770 |
Unconjugated | 0.1 mg |
Each for $499.50
|
|
||||
Description
OPA1 Monoclonal specifically detects OPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
OPA1 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision | |
PBS with 0.02% Sodium Azide | |
4976 | |
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot# O60313] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1284B | |
Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
OPA1 | |
IgG | |
Protein A or G purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title