Learn More
Description
Specifications
Specifications
| Antigen | OPA1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O60313 |
| Gene Alias | FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 |
| Gene Symbols | OPA1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
