Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPA1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 27 publications
$243.50 - $487.50
Specifications
| Antigen | OPA1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11055290T
![]() |
Novus Biologicals
NB11055290SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB11055290
![]() |
Novus Biologicals
NB11055290 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| OPA1 | |
| Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Mitochondrial Fusion Proteins, Neurodegeneration, Neuroscience, Sensory Systems, Vision | |
| PBS with 0.02% Sodium Azide | |
| FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
| OPA1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat and chicken. Predicted to react with primate based on 100% sequence homology. Immunogen sequence has 94% homology to Zebrafish. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Chicken | |
| O60313 | |
| 4976 | |
| A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 111 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title