Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPA1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB354917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354917 Supplier Novus Biologicals Supplier No. NB11055290PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OPA1 Polyclonal antibody specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot

Specifications

Antigen OPA1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation PBS
Gene Alias FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 4976
Target Species Human, Mouse, Rat, Pig, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.