Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPA3 Antibody (OTI3G4), PE, Novus Biologicals™
SDP

Catalog No. NB302168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302168 Supplier Novus Biologicals Supplier No. NBP273147PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OPA3 Monoclonal antibody specifically detects OPA3 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen OPA3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G4
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias FLJ22187, FLJ25932, MGA3Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'), MGC75494, optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia), optic atrophy 3 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human OPA3(NP_079412) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 80207
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.