Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPAL1 Antibody (OPAL1-01), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB002493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002493 Supplier Novus Biologicals Supplier No. NBP262210AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OPAL1 Monoclonal specifically detects OPAL1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen OPAL1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OPAL1-01
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chromosome 10 open reading frame 26, FLJ20154, FLJ20367, MGC125648, OPA1L, OPAL1, outcome predictor in acute leukemia 1
Gene Symbols WBP1L
Host Species Mouse
Immunogen Recombinant fragment of human OPAL1 (amino acids 152-342)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54838
Test Specificity The antibody HCG-61 reacts with beta-subunit of human chorionic gonadotropin, a 40 kDa hormone. hCG belongs to the glycoprotein hormone family together with lutropin (LH), follitropin (FSH) and thyrotropin (TSH). hCG is synthesised by trophoblastic cells of the placenta during pregnancy and stimulates the growth of corpus luteum. Following cross-reactivity (%) was determined by classic double-antibody RIA using unlabelled hormones: beta-hCG (77), alpha-hCG (1.3), hLH (0.86), hFSH (< 0.5), hTSH (< 0.5).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.