Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OPC 21268

Click to view available options
Quantity:
10 mg
50 mg
Description
Nonpeptide vasopressin V1 receptor antagonist. Displays greater affinity for rat V1 than human V1 (Ki values are 25 and 8800 nM respectively). Exhibits 1000-fold selectivity for V1 receptors over V2 receptors. Orally active.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 449.54 |
Purity | >98% |
Molecular Weight (g/mol) | 449.551 |
Chemical Name or Material | OPC 21268 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction