Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Open-Finger Shock Absorbing Glove Liners

Catalog No. p-7214843 Shop All Impacto Protective Products Products
Encompass
Click to view available options
Size:
Small
Medium
Hand:
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding throughout the glove liner offers protection from shock and impact. Use under work gloves in confined spaces to protect the back of the hand.

  • Breathable Nylon Lycra™ is designed to be worn inside work gloves
  • Padded with 3/16 in. impact and shock absorbing VEP pad
  • Open finger design provides tactile feedback
  • Ideal for use with hand tools, or in confined space applications such as mining

Specifications

Color Black
Hand Right
Material Nylon Lycra
Product Type Glove Liners
Size Small
Finger Style Three Quarter
For Use With (Application) Manufacturing Tools, and Confined Spaces
Glove Cuff Style Open
Series 507-01
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.