Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Optineurin Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB328997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328997 Supplier Novus Biologicals Supplier No. NBP300298MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Optineurin Polyclonal antibody specifically detects Optineurin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Optineurin
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Optineurin (Accession#: NP_068815.2; Met1-Ile577)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10133
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.