Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ DdeI (HpyF31), Fisher BioReagents™

Catalog No. p-4362813 Shop All Fisherbrand Products
Click to view available options
Quantity:
1000 U
Product Type:
DdeI (HpyF31)

Catalog No. BP8060-5

5'...C^T N A G...3'
3'...G A N T^C...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
  • Enzyme Activity in OPTIZYME buffers:
    • Buffer 1: 20 - 50%
    • Buffer 2: 20 - 50%
    • Buffer 3: 20 - 50%
    • Buffer 4: 100%
    • Buffer 5: 20 - 50%

    Storage Buffer:
    • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycero
    l
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digesiton with DdeI.
  • Compatible Ends: BbvCI, Bpu10I, Bpu1102I, Eco81I
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: Never overlaps - no effect
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - effect not determined
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Components 25 to 50% Water, >50% Glycerin
Incubator Temperature 37°C
pH 7.4
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digesiton with DdeI
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Content And Storage Keep container tightly closed
Quantity 1000 U
Cut Site C.TNAG
Product Type DdeI (HpyF31)
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.