Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ Terminal Deoxynucleotidyl Transferase, Fisher BioReagents™

Catalog No. BP81035 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP81035 1500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP81035

missing translation for 'validMainframeMsg'

OPTIZYME™ TdT is a template-independent DNA polymerase that catalyzes the repetitive addition of deoxyribonucleotides to the 3'-OH group of DNA

Applications and Features:

  • OPTIZYME TdT produces synthetic homo- and heteropolymers
  • Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus
  • Oligodeoxyribonucleotide and DNA labeling
  • 5'-Rapid Amplification of cDNA Ends (5'-RACE)
  • In situ localization of apoptosis
  • Incorporation of modified nucleotides (e.g., fluorescein-, biotin-, aminoallyl-labeled nucleotides)

Supplied with:
5X OPTIZYME TdT Buffer: 1M potassium cacodylate, 125mM Tris, 0.05% (v/v) Triton X-100, 5mM CoCl2 (pH 7.2 at 25°C)

Source: E. Coli cells with a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase

Quality Control:
Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Color Colorless
pH 7.5
For Use With (Application) Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus, Oligodeoxyribonucleotide and DNA labeling, 5'-rapid amplification of cDNA Ends, in-situ localization of apoptosis incorporation of modified Nucleotides
Storage Buffer OPTIZYME™ TdT Buffer 5X : Potassium cacodylate 1M, Tris 125mM, Triton™ X-100 0.05%, CoCl2 5mM (7.2 pH at 25°C)
Content And Storage -20°C
Source E. coli cells with a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
Quantity 1500 U
Product Type Terminal Deoxynucleotidyl Transferase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.