Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ XbaI, Fisher BioReagents™

Catalog No. p-4362858 Shop All Fisherbrand Products
Click to view available options
Quantity:
2000 U
Concentration:
10 U/μL

5'...T^C T A G A...3'
3'...A G A T C^T...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 - 100%
  • Buffer 2: 50 - 100%
  • Buffer 3: 20 - 50%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 7mM 2-mercaptoethanol, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Ligation and Re-cleavage:
More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with XbaI.
Digestion of Agarose-embedded DNA:
A minimum 5 units of XbaI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Notes:
  • XbaI is blocked by overlapping dam methylation.
  • To avoid dam methylation, use a dam-, dcm- strain.
Compatible Ends: BcuI, Eco130I, NheI, XmaJI
Methylation Effects:
Dam: May overlap - blocked
  • 5'...TCTAGm6A TC...3'
  • 3'...AGATC Tm6AG...5'
  • (Cleavage is blocked)
Dcm: Never overlaps - no effect
CpG: Never overlaps - no effect
EcoKI: Never overlaps - no effect
EcoBI: Never overlaps - no effect

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Components 10X OPTIZYME™ Buffer 4
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Indicates ligation efficiency of DNA fragments generated by digestion with the Restriction Enzyme
Storage Buffer 10mM Tris HCl (pH 7.4), 100mM KCl, 7mM 2-mercaptoethano, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Quantity 2000 U
Cut Site T.CTAGA
Product Type XbaI
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.