Learn More
Description
Specifications
Specifications
| Antigen | OR10G9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Olfactory receptor 10G10, olfactory receptor 10G9, olfactory receptor OR11-272, olfactory receptor, family 10, subfamily G, member 9, OR10G10P |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR10G9 (NP_001001953). Peptide sequence IYLRPGSRDVVDGVVAIFYTVLTPLLNPVVYTLRNKEVKKAVLKLRDKVA |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
