Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR14I1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309860100UL
Description
OR14I1 Polyclonal specifically detects OR14I1 in Human samples. It is validated for Western Blot.Specifications
OR14I1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
olfactory receptor 14I1, Olfactory receptor 5BU1, olfactory receptor, family 14, subfamily I, member 1, olfactory receptor, family 5, subfamily BU, member 1, olfactory receptor, family 5, subfamily BU, member 1 pseudogene, OR5BU1, OR5BU1P | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR14I1. Peptide sequence LGCFILMMISYFQIFSTVLRIPSGQSRAKAFSTCSPQLIVIMLFLTTGLF | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
401994 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction