Learn More
Description
Specifications
Specifications
| Antigen | OR14J1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | bA150A6.4, hs6M1-28, olfactory receptor 14J1, Olfactory receptor 5U1, Olfactory receptor OR6-25, olfactory receptor, family 14, subfamily J, member 1, olfactory receptor, family 5, subfamily U member 1, olfactory receptor, family 5, subfamily U, member 1, OR5U1, OR6-25 |
| Gene Symbols | OR14J1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YSLRNDSMKAALRKMLSKEELPQR |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
