Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR1M1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310247100UL
Description
OR1M1 Polyclonal specifically detects OR1M1 in Mouse samples. It is validated for Western Blot.Specifications
OR1M1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
olfactory receptor 1M1, Olfactory receptor OR19-5, olfactory receptor, family 1, subfamily M, member 1, OR19-5, OR19-6Olfactory receptor 19-6 | |
The immunogen is a synthetic peptide directed towards the C terminal region of mouse OR1M1 (NP_666817.1). Peptide sequence CPSSVRTAVKEKASAVMYTAVTPMLNPFIYSLRNRDLKGALKKIINRKIS | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
125963 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction